missing translation for 'onlineSavingsMsg'
Learn More

1-Iodonaphthalene 97.0+%, TCI America™

Catalog No. I026625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I026625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I026625G Supplier TCI America Supplier No. I026625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-14-2
Molecular Formula C10H7I
Molecular Weight (g/mol) 254.07
MDL Number MFCD00003876
InChI Key NHPPIJMARIVBGU-UHFFFAOYSA-N
Synonym naphthalene, 1-iodo, 1-naphthyl iodide, alpha-iodonaphthalene, iodonaphthalene, 1-iodonaphthalenen, .alpha.-iodonaphthalene, 1-iodo-naphthalene, 1-iodonaphthlen, 1-iodonaphthlene, 1-iodonapthalene
PubChem CID 7004
IUPAC Name 1-iodonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2I

Specifications

Color Brown-Yellow
Boiling Point 305°C
Quantity 25 g
Formula Weight 254.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodonaphthalene