missing translation for 'onlineSavingsMsg'
Learn More

1-Iodonaphthalene, 98%

Catalog No. p-7026578
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2264409 10 g
AAB2264418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2264409 Supplier Thermo Scientific Chemicals Supplier No. B2264409

CAS: 90-14-2 | C10H7I | 254.07 g/mol

1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 90-14-2
Molecular Formula C10H7I
Molecular Weight (g/mol) 254.07
MDL Number MFCD00003876
InChI Key NHPPIJMARIVBGU-UHFFFAOYSA-N
Synonym naphthalene, 1-iodo, 1-naphthyl iodide, alpha-iodonaphthalene, iodonaphthalene, 1-iodonaphthalenen, .alpha.-iodonaphthalene, 1-iodo-naphthalene, 1-iodonaphthlen, 1-iodonaphthlene, 1-iodonapthalene
PubChem CID 7004
IUPAC Name 1-iodonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2I

Specifications

Melting Point 4°C
Density 1.738
Boiling Point 161°C to 162°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.701
Quantity 10 g
Beilstein 1906415
Sensitivity Light Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 254.07
Percent Purity 98%
Chemical Name or Material 1-Iodonaphthalene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-968-9
TSCA Yes

RUO – Research Use Only