missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexadecane 97.0+%, TCI America™

Catalog No. I040825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I040825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I040825G Supplier TCI America Supplier No. I040825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 544-77-4
Molecular Formula C16H33I
Molecular Weight (g/mol) 352.344
MDL Number MFCD00001089
InChI Key KMWHQYDMBYABKL-UHFFFAOYSA-N
Synonym cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide
PubChem CID 11007
IUPAC Name 1-iodohexadecane
SMILES CCCCCCCCCCCCCCCCI

Specifications

Color White-Yellow
Boiling Point 207°C
Quantity 25 g
Formula Weight 352.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Iodohexadecane