missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexadecane, 98%, stab. with copper

Catalog No. AAA1297714
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1297714 25 g
AAA1297722 100 g
AAA1297730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1297714 Supplier Thermo Scientific Chemicals Supplier No. A1297714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 544-77-4 | C16H33I | 352.344 g/mol

1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from strong oxidizing agents, strong bases. Store it at a temperature of 15 - 30°C. Stable under normal temperatures and pressures.

Chemical Identifiers

CAS 544-77-4
Molecular Formula C16H33I
Molecular Weight (g/mol) 352.344
MDL Number MFCD00001089
InChI Key KMWHQYDMBYABKL-UHFFFAOYSA-N
Synonym cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide
PubChem CID 11007
IUPAC Name 1-iodohexadecane
SMILES CCCCCCCCCCCCCCCCI

Specifications

Melting Point 21°C to 23°C
Density 1.121
Boiling Point 206°C to 207°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.482
Linear Formula CH3(CH2)15I
Quantity 25 g
Beilstein 1748533
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 352.34
Percent Purity 98%
Chemical Name or Material 1-Iodohexadecane, stabilized with copper
Show More Show Less
EINECSNumber 208-879-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only