missing translation for 'onlineSavingsMsg'
Learn More

1-Iodoheptane, 98%, stab. with copper

Catalog No. p-7027449
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2420618 50 g
AAB2420630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2420618 Supplier Thermo Scientific Chemicals Supplier No. B2420618

CAS: 4282-40-0 | C7H15I | 226.101 g/mol

1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4282-40-0
Molecular Formula C7H15I
Molecular Weight (g/mol) 226.101
MDL Number MFCD00001104
InChI Key LMHCYRULPLGEEZ-UHFFFAOYSA-N
Synonym heptyl iodide, n-heptyl iodide, heptane, 1-iodo, iodoheptane, n-heptyliodide, iodo heptane, 1-iodanylheptane, 1-iodomeptane, 1-iodo heptane, 1-iodo-heptane
PubChem CID 20274
IUPAC Name 1-iodoheptane
SMILES CCCCCCCI

Specifications

Melting Point -48°C
Density 1.379
Boiling Point 203°C to 205°C
Flash Point 78°C (174°F)
Refractive Index 1.49
Quantity 50 g
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 226.1
Percent Purity 98%
Chemical Name or Material 1-Iodoheptane, stabilized with copper
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 224-285-8
RTECSNumber MJ1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only