missing translation for 'onlineSavingsMsg'
Learn More

1-Iodododecane, 98%, stabilized

Catalog No. AC250091000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250091000 100 mL Glass bottle
AC250090250 25 mL Glass bottle
AC250095000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC250091000 Supplier Thermo Scientific Chemicals Supplier No. 250091000
Only null left

CAS: 4292-19-7 | C12H25I | 296.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4292-19-7
Molecular Formula C12H25I
Molecular Weight (g/mol) 296.24
MDL Number MFCD00001088
InChI Key GCDPERPXPREHJF-UHFFFAOYSA-N
Synonym n-dodecyl iodide, dodecyl iodide, lauryl iodide, dodecane, 1-iodo, dodecane,1-iodo, iodododecane, iodo-dodecane, 1-iodanyldodecane, pubchem3943, n-iodododecane
PubChem CID 20282
IUPAC Name 1-iodododecane
SMILES CCCCCCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -3.0°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 159.0°C to 160.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.4850
Linear Formula CH3(CH2)11I
Quantity 100 mL
Beilstein 01,67
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 296.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodododecane, Stabilized with 500ppm copper
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-293-1

RUO – Research Use Only