Learn More
1-Iodododecane, 98%, stab. with copper
CAS: 4292-19-7 | C12H25I | 296.24 g/mol
Supplier: Thermo Scientific Chemicals A1190922
Description
1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
Specifications
| 1-Iodododecane | |
| 4292-19-7 | |
| 1.201 | |
| >110°C (230°F) | |
| 1.4844 | |
| 100 g | |
| Light sensitive | |
| Not miscible or difficult to mix in water. | |
| CCCCCCCCCCCCI | |
| 296.24 | |
| 296.24 |
| Stabilized with copper | |
| -3°C | |
| 128°C to 130°C (11 mmHg) | |
| C12H25I | |
| MFCD00001088 | |
| 1738943 | |
| n-dodecyl iodide, dodecyl iodide, lauryl iodide, dodecane, 1-iodo, dodecane,1-iodo, iodododecane, iodo-dodecane, 1-iodanyldodecane, pubchem3943, n-iodododecane | |
| GCDPERPXPREHJF-UHFFFAOYSA-N | |
| 1-iodododecane | |
| 20282 | |
| 98% |
Chemical Identifiers
| 4292-19-7 | |
| 296.24 | |
| GCDPERPXPREHJF-UHFFFAOYSA-N | |
| 1-iodododecane |
| C12H25I | |
| MFCD00001088 | |
| 20282 | |
| CCCCCCCCCCCCI |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-293-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only