missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iododecane (stabilized with Copper chip) 96.0+%, TCI America™
Supplier: TCI America I0494250G
Specifications
| 1-Iododecane (stabilized with Copper chip) | |
| Red | |
| C10H21I | |
| 250 g | |
| SKIDNYUZJPMKFC-UHFFFAOYSA-N | |
| 1-iododecane | |
| 16314 | |
| ≥96.0% (GC) |
| 2050-77-3 | |
| 132°C | |
| MFCD00001085 | |
| decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942 | |
| CCCCCCCCCCI | |
| 268.18 | |
| 268.18 | |
| Liquid |
Chemical Identifiers
| 2050-77-3 | |
| 268.18 | |
| SKIDNYUZJPMKFC-UHFFFAOYSA-N | |
| 16314 | |
| CCCCCCCCCCI |
| C10H21I | |
| MFCD00001085 | |
| decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942 | |
| 1-iododecane |
Safety and Handling
RTECSNumber : HD8705000
TSCA : Yes