missing translation for 'onlineSavingsMsg'
Learn More

1-Iododecane (stabilized with Copper chip) 96.0+%, TCI America™

Catalog No. I049425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I049425G 25 g
I0494250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I049425G Supplier TCI America Supplier No. I049425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2050-77-3
Molecular Formula C10H21I
Molecular Weight (g/mol) 268.18
MDL Number MFCD00001085
InChI Key SKIDNYUZJPMKFC-UHFFFAOYSA-N
Synonym decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942
PubChem CID 16314
IUPAC Name 1-iododecane
SMILES CCCCCCCCCCI

Specifications

Color Red
Boiling Point 132°C
Quantity 25 g
Formula Weight 268.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iododecane (stabilized with Copper chip)
RTECSNumber HD8705000
TSCA Yes