missing translation for 'onlineSavingsMsg'
Learn More

1-Iododecane, 98%, stab. with copper

Catalog No. AAA1617330
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1617330 250 g
AAA1617314 25 g
AAA1617318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1617330 Supplier Thermo Scientific Chemicals Supplier No. A1617330

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2050-77-3 | C10H21I | 268.18 g/mol

1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases. Keep the container tightly closed.

Chemical Identifiers

CAS 2050-77-3
Molecular Formula C10H21I
Molecular Weight (g/mol) 268.18
MDL Number MFCD00001085
InChI Key SKIDNYUZJPMKFC-UHFFFAOYSA-N
PubChem CID 16314
IUPAC Name 1-iododecane
SMILES CCCCCCCCCCI

Specifications

Melting Point -16°C
Density 1.257
Boiling Point 264°C
Flash Point 96°C (204°F)
Refractive Index 1.485
Linear Formula CH3(CH2)9I
Quantity 250 g
Beilstein 1735228
Sensitivity Light sensitive
Synonym decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942
Solubility Information Immiscible with water.
Formula Weight 268.18
Percent Purity 98%
Chemical Name or Material 1-Iododecane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-104-1
RTECSNumber HD8705000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only