missing translation for 'onlineSavingsMsg'
Learn More

1-Iodobutane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I005525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I005525G 25 g
I0055500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I005525G Supplier TCI America Supplier No. I005525G

Chemical Identifiers

CAS 542-69-8
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001098
InChI Key KMGBZBJJOKUPIA-UHFFFAOYSA-N
Synonym butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide
PubChem CID 10962
IUPAC Name 1-iodobutane
SMILES CCCCI

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 1993
Formula Weight 184.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodobutane (stabilized with Copper chip)
EINECSNumber (2)-0063
RTECSNumber EK4400000
TSCA Yes