missing translation for 'onlineSavingsMsg'
Learn More

1-Iodobutane, 99%, stab. with copper

Catalog No. AAA1234036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1234036 500 g
AAA1234022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1234036 Supplier Thermo Scientific Chemicals Supplier No. A1234036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 542-69-8 | C4H9I | 184.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Iodobutane was used in the synthesis of S-alkylated 5-(2-,3- and 4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol. Used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep away from sources of ignition. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances like Strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 542-69-8
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001098
InChI Key KMGBZBJJOKUPIA-UHFFFAOYSA-N
Synonym butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide
PubChem CID 10962
IUPAC Name 1-iodobutane
SMILES CCCCI

Specifications

Melting Point -103°C
Density 1.616
Boiling Point 129°C to 131°C
Flash Point 33°C (91°F)
Assay Percent Range 99%, Stabilized with Copper
Refractive Index 1.499
Linear Formula CH3(CH2)3I
Quantity 500 g
UN Number UN1993
Beilstein 1420755
Sensitivity Light sensitive
Merck Index 14,1573
Solubility Information Insoluble in water.
Formula Weight 184.02
Percent Purity 99%
Chemical Name or Material 1-Iodobutane, Stabilized with copper
Show More Show Less
Hazard Category H226-H315-H319-H331-H335
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 208-824-4
RTECSNumber EK4400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only