missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-methylbutane, 97%, stab. with copper

Catalog No. AAL0519409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0519409 10 g
AAL0519418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0519409 Supplier Thermo Scientific Chemicals Supplier No. L0519409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 541-28-6 | C5H11I | 198.047 g/mol

1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.

Solubility
Insoluble in water. Soluble in most common organic solvents.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 541-28-6
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.047
MDL Number MFCD00039414
InChI Key BUZZUHJODKQYTF-UHFFFAOYSA-N
Synonym isoamyl iodide, isopentyl iodide, butane, 1-iodo-3-methyl, 3-methylbutyliodide, 1-iodo-3-methyl-butane, 1-jod-3-methylbutan, iso-amyliodide, unii-e1e5du187l, 1-jod-3-methylbutan czech, 1-iodo-3-methyl butane
PubChem CID 10924
IUPAC Name 1-iodo-3-methylbutane
SMILES CC(C)CCI

Specifications

Density 1.515
Boiling Point 146°C to 148°C
Flash Point 40°C (104°F)
Refractive Index 1.492
Quantity 10 g
UN Number UN1993
Beilstein 1730990
Sensitivity Light sensitive
Merck Index 14,5120
Solubility Information Insoluble in water. Soluble in most common organic solvents.
Formula Weight 198.05
Percent Purity 97%
Chemical Name or Material 1-Iodo-3-methylbutane, stabilized with copper
Show More Show Less
Hazard Category H226-H302
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 208-773-8
RTECSNumber EK4412000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only