missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-1-propyne 97.0+%, TCI America™

Catalog No. P10651G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10651G Supplier TCI America Supplier No. P10651G
Only null left

Chemical Identifiers

CAS 624-66-8
Molecular Formula C3H3I
Molecular Weight (g/mol) 165.961
MDL Number MFCD00059101
InChI Key JHUPGXNKUPOSIE-UHFFFAOYSA-N
Synonym 1-Propyn-1-yl Iodide
PubChem CID 136437
IUPAC Name 1-iodoprop-1-yne
SMILES CC#CI

Specifications

Boiling Point 110°C
Quantity 1 g
UN Number 1993
Formula Weight 165.96
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-1-propyne
TSCA No
Recommended Storage Refrigerator