missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Indolyl)acetic Acid 97.0+%, TCI America™
$85.75 - $212.51
Chemical Identifiers
| CAS | 24297-59-4 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.187 |
| MDL Number | MFCD00047262 |
| InChI Key | WQJFIWXYPKYBTO-UHFFFAOYSA-N |
| Synonym | 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid |
| PubChem CID | 90448 |
| ChEBI | CHEBI:72814 |
| IUPAC Name | 2-indol-1-ylacetic acid |
| SMILES | C1=CC=C2C(=C1)C=CN2CC(=O)O |
Chemical Identifiers
| 24297-59-4 | |
| 175.187 | |
| WQJFIWXYPKYBTO-UHFFFAOYSA-N | |
| 90448 | |
| 2-indol-1-ylacetic acid |
| C10H9NO2 | |
| MFCD00047262 | |
| 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid | |
| CHEBI:72814 | |
| C1=CC=C2C(=C1)C=CN2CC(=O)O |
Specifications
| 24297-59-4 | |
| C10H9NO2 | |
| 1 g | |
| WQJFIWXYPKYBTO-UHFFFAOYSA-N | |
| 2-indol-1-ylacetic acid | |
| 90448 | |
| 175.19 | |
| Crystalline Powder |
| 170°C | |
| MFCD00047262 | |
| 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid | |
| C1=CC=C2C(=C1)C=CN2CC(=O)O | |
| 175.187 | |
| CHEBI:72814 | |
| ≥97.0% (T) | |
| (1-Indolyl)acetic Acid |
Safety and Handling
TSCA : No