missing translation for 'onlineSavingsMsg'
Learn More

(1-Indolyl)acetic Acid 97.0+%, TCI America™

Catalog No. I09325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09325G 5 g
I09321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09325G Supplier TCI America Supplier No. I09325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24297-59-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00047262
InChI Key WQJFIWXYPKYBTO-UHFFFAOYSA-N
Synonym 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid
PubChem CID 90448
ChEBI CHEBI:72814
IUPAC Name 2-indol-1-ylacetic acid
SMILES C1=CC=C2C(=C1)C=CN2CC(=O)O

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1-Indolyl)acetic Acid