missing translation for 'onlineSavingsMsg'
Learn More

1-Indoleacetic acid, 95%

Catalog No. AAH63935MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63935MD 250 mg
AAH6393503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63935MD Supplier Thermo Scientific Chemicals Supplier No. H63935MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24297-59-4 | C10H9NO2 | 175.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24297-59-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00047262
InChI Key WQJFIWXYPKYBTO-UHFFFAOYSA-N
Synonym 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid
PubChem CID 90448
ChEBI CHEBI:72814
IUPAC Name 2-indol-1-ylacetic acid
SMILES C1=CC=C2C(=C1)C=CN2CC(=O)O

Specifications

Quantity 250 mg
Beilstein 142924
Formula Weight 175.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Indoleacetic acid
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only