missing translation for 'onlineSavingsMsg'
Learn More

1-Imidazoleacetic Acid 98.0+%, TCI America™

Catalog No. I06705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06705G 5 g
I067025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06705G Supplier TCI America Supplier No. I06705G

Chemical Identifiers

CAS 22884-10-2
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD01721800
InChI Key QAFBDRSXXHEXGB-UHFFFAOYSA-N
Synonym imidazol-1-yl-acetic acid, 2-1h-imidazol-1-yl acetic acid, 1h-imidazole-1-acetic acid, 1h-imidazol-1-ylacetic acid, imidazole-1-acetic acid, 1-imidazolyl acetic acid, 1-imidazoleacetic acid, imidazol-1-ylacetic acid, imidazol-1-yl acetic acid, 1-imidazolylacetic acid
PubChem CID 656129
ChEBI CHEBI:70801
IUPAC Name 2-imidazol-1-ylacetic acid
SMILES C1=CN(C=N1)CC(=O)O

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 5 g
Formula Weight 126.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Imidazoleacetic Acid