missing translation for 'onlineSavingsMsg'
Learn More

1-(Hydroxymethyl)-5,5-dimethylhydantoin 98.0+%, TCI America™

Catalog No. M0668500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0668500G 500 g
M066825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0668500G Supplier TCI America Supplier No. M0668500G

Chemical Identifiers

CAS 116-25-6
Molecular Formula C6H10N2O3
Molecular Weight (g/mol) 158.16
MDL Number MFCD00020866
InChI Key SIQZJFKTROUNPI-UHFFFAOYSA-N
Synonym 1-hydroxymethyl-5,5-dimethylhydantoin, mdm hydantoin, glycoserve, mdmh, 1-hydroxymethyl-5,5-dimethylimidazolidine-2,4-dione, mdm hydantoin van, monomethyloldimethylhydantoin, 1-monomethylol-5,5-dimethylhydantoin, methylol dimethylhydantoin
PubChem CID 67000
IUPAC Name 1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)N(CO)C(=O)NC1=O

Specifications

Color White
Quantity 500 g
Formula Weight 158.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Hydroxymethyl)-5,5-dimethylhydantoin
EINECSNumber (5)-3543
TSCA Yes