missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Hydroxymethyl)-5,5-dimethylhydantoin 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 158.16 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-(Hydroxymethyl)-5,5-dimethylhydantoin |
Safety and Handling
| EINECSNumber | (5)-3543 |
| TSCA | Yes |