missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxyethylferrocene 95.0+%, TCI America™
$59.65 - $244.19
Chemical Identifiers
| CAS | 1277-49-2 |
|---|---|
| Molecular Formula | C12H14FeO |
| Molecular Weight (g/mol) | 230.09 |
| MDL Number | MFCD00013752 |
| InChI Key | KGIBJAFJTDOHBV-LURJTMIESA-N |
| Synonym | 1-Ferrocenylethanol |
| IUPAC Name | (±)-1-Ferrocenylethanol |
| SMILES | [Fe].c1cccc1.CC(O)c1cccc1 |
Chemical Identifiers
| 1277-49-2 | |
| 230.09 | |
| KGIBJAFJTDOHBV-LURJTMIESA-N | |
| (±)-1-Ferrocenylethanol |
| C12H14FeO | |
| MFCD00013752 | |
| 1-Ferrocenylethanol | |
| [Fe].c1cccc1.CC(O)c1cccc1 |
Specifications
| 1277-49-2 | |
| C12H14FeO | |
| 1 g | |
| KGIBJAFJTDOHBV-LURJTMIESA-N | |
| (±)-1-Ferrocenylethanol | |
| 230.09 | |
| Crystalline Powder |
| 78°C | |
| MFCD00013752 | |
| 1-Ferrocenylethanol | |
| [Fe].c1cccc1.CC(O)c1cccc1 | |
| 230.09 | |
| ≥95.0% (T) | |
| 1-Hydroxyethylferrocene |
Safety and Handling
RTECSNumber : LK0730000
TSCA : No