missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyethylethoxypiperazine, 95%

Catalog No. AC259111000
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259111000 100 mL Glass bottle
AC259110050 5 mL Glass bottle
AC259110250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC259111000 Supplier Thermo Scientific Chemicals Supplier No. 259111000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13349-82-1 | C8H18N2O2 | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13349-82-1
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.24
MDL Number MFCD00074931
InChI Key FLNQAPQQAZVRDA-UHFFFAOYSA-N
Synonym 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
PubChem CID 139436
IUPAC Name 2-(2-piperazin-1-ylethoxy)ethanol
SMILES C1CN(CCN1)CCOCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown-Yellow to Colorless
Density 1.0100g/mL
Boiling Point 150.0°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Quantity 100 mL
Specific Gravity 1.01
Formula Weight 174.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Hydroxyethylethoxypiperazine
Show More Show Less

RUO – Research Use Only