missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyethane-1,1-diphosphonic Acid (ca. 60% in Water, ca. 4.2mol/L), TCI America™

Catalog No. p-7130621
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H058725G 25 g
H0587500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H058725G Supplier TCI America Supplier No. H058725G
Only null left

Chemical Identifiers

CAS 2809-21-4
Molecular Formula C2H8O7P2
Molecular Weight (g/mol) 206.027
MDL Number MFCD00070585
InChI Key DBVJJBKOTRCVKF-UHFFFAOYSA-N
Synonym etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl
PubChem CID 3305
ChEBI CHEBI:4907
IUPAC Name (1-hydroxy-1-phosphonoethyl)phosphonic acid
SMILES CC(O)(P(=O)(O)O)P(=O)(O)O

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 105°C
Quantity 25 g
UN Number 3265
Formula Weight 206.03
Physical Form Liquid
Chemical Name or Material 1-Hydroxyethane-1,1-diphosphonic Acid (ca. 60% in Water, ca. 4.2mol/L)
EINECSNumber (2)-2936
RTECSNumber SZ8562100
TSCA Yes