missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxycyclohexyl Phenyl Ketone 98.0+%, TCI America™

Catalog No. p-7130637
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H061725G 25 g
H0617100G 100 g
H0617500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H061725G Supplier TCI America Supplier No. H061725G
Only null left

Chemical Identifiers

CAS 947-19-3
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00059561
InChI Key QNODIIQQMGDSEF-UHFFFAOYSA-N
Synonym 1-hydroxycyclohexyl phenyl ketone, 1-hydroxycyclohexyl phenyl methanone, 1-benzoylcyclohexanol, hydroxycyclohexyl phenyl ketone, 1-benzoylcyclohexan-1-ol, irgacure 184, methanone, 1-hydroxycyclohexyl phenyl, unii-e7jvn2243x, 1-hydroxycyclohexyl-phenylmethanone, 1-cyclohexylhydroxyphenylketone
PubChem CID 70355
IUPAC Name (1-hydroxycyclohexyl)-phenylmethanone
SMILES C1CCC(CC1)(C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 25 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxycyclohexyl Phenyl Ketone
EINECSNumber (4)-1397
TSCA Yes