missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxycyclohexyl Phenyl Ketone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 49°C |
| Color | White-Yellow |
| Quantity | 100 g |
| Formula Weight | 204.27 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Hydroxycyclohexyl Phenyl Ketone |
Safety and Handling
| EINECSNumber | (4)-1397 |
| TSCA | Yes |