missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-1-cyclopropanecarboxylic acid, 97%

Catalog No. AC301430010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301430010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC301430010 Supplier Thermo Scientific Chemicals Supplier No. 301430010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17994-25-1 | C4H5O3 | 101.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17994-25-1
Molecular Formula C4H5O3
Molecular Weight (g/mol) 101.08
MDL Number MFCD00010797
InChI Key GQXURJDNDYACGE-UHFFFAOYSA-M
Synonym 1-hydroxy-1-cyclopropanecarboxylic acid, 1-hydroxycyclopropanecarboxylic acid, 1-hydroxy-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-hydroxy, 1-hydroxy-1-cyclopropyl carboxylic acid, pubchem12465, acmc-20a5ho, 1-carboxy-1-hydroxycyclopropane, 1-hydroxycyclopropanecaboxylic acid, 1-hydroxycyclo-propanecarboxylic acid
PubChem CID 2733160
IUPAC Name 1-hydroxycyclopropane-1-carboxylic acid
SMILES OC1(CC1)C([O-])=O

Specifications

Melting Point 108.0°C to 110.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 102.09
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Hydroxy-1-cyclopropanecarboxylic acid
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only