missing translation for 'onlineSavingsMsg'
Learn More

ENG Scientific Bone Marrow Stain Kit for Iron, Solution II - Hydrochloric Acid (1%) DFS Item

Bone Marrow Stain Kit for Iron for Iron Prussion reaction. Hemosiderin and ferritin stain blue and is usually reported as either negative or 1 to 5. Iron in hemoglobin is unstained.

Supplier:  ENG Scientific ES3452

Catalog No. ES3452

Only null left
Explore available promotions

Description

Description

  • Bone Marrow Stain Kit for Iron; Solution II - Hydrochloric Acid (1%)
  • For Iron (Prussian blue reaction)
  • Hemosiderin and ferritin stain blue and is usually reported as either negative or 1 to 5
  • Iron in hemoglobin is unstained
Specifications

Specifications

Hydochloric Acid
1%
Bottles
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For in vitro diagnostic use only.