missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyn-3-ol 95.0+%, TCI America™

Catalog No. h01415ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01415ML 5 mL
H014125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01415ML Supplier TCI America Supplier No. H01415ML
Only null left

Chemical Identifiers

CAS 105-31-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00014408
InChI Key LTFTWJYRQNTCHI-UHFFFAOYSA-N
Synonym 1-hexyn-3-ol, propylethinylcarbinol, 1-hexyne-3-ol, 3-hydroxy-1-hexyne, 1-propylpropargyl alcohol, 1-hexine-3-ol, acmc-2098fm, dsstox_cid_30088, dsstox_gsid_51536, 4-01-00-02234 beilstein handbook reference
PubChem CID 7746
IUPAC Name hex-1-yn-3-ol
SMILES CCCC(C#C)O

Specifications

Color Yellow
Boiling Point 141°C
Quantity 5 mL
UN Number 2929
Formula Weight 98.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexyn-3-ol
EINECSNumber (2)-2397
RTECSNumber MR0181000
TSCA Yes