missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexylboronic acid pinacol ester, 98%
CAS: 86308-26-1 | C12H25BO2 | 212.14 g/mol
$76.96 - $873.82
Chemical Identifiers
| CAS | 86308-26-1 |
|---|---|
| Molecular Formula | C12H25BO2 |
| Molecular Weight (g/mol) | 212.14 |
| MDL Number | MFCD08457665 |
| InChI Key | LDPMCSWZEGKWLC-UHFFFAOYSA-N |
| Synonym | hexylboronic acid pinacol ester, 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 1,3,2-dioxaborolane, 2-hexyl-4,4,5,5-tetramethyl |
| PubChem CID | 10921795 |
| IUPAC Name | 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2783103
|
Thermo Scientific Chemicals
H2783103 |
1 g |
Each for $76.96
|
|
|||||
|
AAH2783106
|
Thermo Scientific Chemicals
H2783106 |
5 g |
Each for $268.33
|
|
|||||
|
AAH2783114
|
Thermo Scientific Chemicals
H2783114 |
25 g |
Each for $873.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 86308-26-1 | |
| 212.14 | |
| LDPMCSWZEGKWLC-UHFFFAOYSA-N | |
| 10921795 | |
| B1(OC(C(O1)(C)C)(C)C)CCCCCC |
| C12H25BO2 | |
| MFCD08457665 | |
| hexylboronic acid pinacol ester, 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 1,3,2-dioxaborolane, 2-hexyl-4,4,5,5-tetramethyl | |
| 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
| 86308-26-1 | |
| 84°C (183°F) | |
| 1.426 | |
| 1 g | |
| LDPMCSWZEGKWLC-UHFFFAOYSA-N | |
| 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 10921795 | |
| 98% |
| 1.167 | |
| C12H25BO2 | |
| MFCD08457665 | |
| hexylboronic acid pinacol ester, 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 1,3,2-dioxaborolane, 2-hexyl-4,4,5,5-tetramethyl | |
| B1(OC(C(O1)(C)C)(C)C)CCCCCC | |
| 212.14 | |
| 212.14 | |
| 1-Hexylboronic acid pinacol ester |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only