missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylboronic acid, 97%

Catalog No. p-7026448
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2244688 2.5 g
AAB2244609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2244688 Supplier Thermo Scientific Chemicals Supplier No. B2244688
Only null left

CAS: 16343-08-1 | C6H15BO2 | 129.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16343-08-1
Molecular Formula C6H15BO2
Molecular Weight (g/mol) 129.994
MDL Number MFCD01074641
InChI Key CXSYDLCMCLCOCA-UHFFFAOYSA-N
Synonym n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid, boronic acid, hexyl, b-hexyl-boronic acid, n-hexylboronicacid, 1-hexylboronic acid c6-b oh 2
PubChem CID 351064
IUPAC Name hexylboronic acid
SMILES B(CCCCCC)(O)O

Specifications

Melting Point 85°C to 90°C
Quantity 2.5 g
Beilstein 1736080
Sensitivity Air Sensitive
Formula Weight 130
Percent Purity 97%
Chemical Name or Material 1-Hexylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only