missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexyl)trimethylammonium bromide, 98%

Catalog No. p-7044879
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0859406 5 g
AAL0859414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0859406 Supplier Thermo Scientific Chemicals Supplier No. L0859406
Only null left

CAS: 2650-53-5 | C9H22BrN | 224.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2650-53-5
Molecular Formula C9H22BrN
Molecular Weight (g/mol) 224.186
MDL Number MFCD00051872
InChI Key JYVPKRHOTGQJSE-UHFFFAOYSA-M
Synonym hexyltrimethylammonium bromide, hexyltrimethylazanium bromide, acmc-209grw, hexyltrimethylammoniumbromide, n-hexyltrimethylammonium bromide, n,n,n,n-hexyltrimethylammonium bromide, 1-hexanaminium,n,n-trimethyl-, bromide, 1-hexyl trimethylammonium bromide, 1-hexanaminium,n,n,n-trimethyl-, bromide 1:1
PubChem CID 10059492
IUPAC Name hexyl(trimethyl)azanium;bromide
SMILES CCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 177°C to 180°C
Quantity 5 g
Beilstein 3913229
Sensitivity Hygroscopic
Formula Weight 224.18
Percent Purity 98%
Chemical Name or Material (1-Hexyl)trimethylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only