missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Hexafluorophosphate 98.0+%, TCI America™

Supplier:  TCI America H10985G

 View more versions of this product

Catalog No. H10985G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Hexyl-3-methylimidazolium Hexafluorophosphate
Yellow
MFCD03093296
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
312.24
312.24
Liquid
304680-35-1
C10H19F6N2P
5 g
YPWSSSRXUOQNMQ-UHFFFAOYSA-N
1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
2734175
≥98.0% (N,T)

Chemical Identifiers

304680-35-1
312.24
YPWSSSRXUOQNMQ-UHFFFAOYSA-N
2734175
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
C10H19F6N2P
MFCD03093296
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions