missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexyl-3-methylimidazolium Bromide 98.0+%, TCI America™
Supplier: TCI America H122725G
Specifications
| 1-Hexyl-3-methylimidazolium Bromide | |
| 395°C | |
| MFCD03427612 | |
| BGSUDDILQRFOKZ-UHFFFAOYSA-M | |
| 1-hexyl-3-methylimidazol-3-ium;bromide | |
| 2734237 | |
| ≥98.0% (HPLC,T) |
| 85100-78-3 | |
| C10H19BrN2 | |
| 25 g | |
| CCCCCCN1C=C[N+](=C1)C.[Br-] | |
| 247.18 | |
| 247.18 | |
| Liquid |
Chemical Identifiers
| 85100-78-3 | |
| 247.18 | |
| BGSUDDILQRFOKZ-UHFFFAOYSA-M | |
| 1-hexyl-3-methylimidazol-3-ium;bromide |
| C10H19BrN2 | |
| MFCD03427612 | |
| 2734237 | |
| CCCCCCN1C=C[N+](=C1)C.[Br-] |
Safety and Handling
TSCA : No