missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene 97.0+%, TCI America™

Catalog No. p-7130356
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H012125ML 25 mL
H0121500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H012125ML Supplier TCI America Supplier No. H012125ML
Only null left

Chemical Identifiers

CAS 592-41-6
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009505
InChI Key LIKMAJRDDDTEIG-UHFFFAOYSA-N
Synonym 1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
PubChem CID 11597
ChEBI CHEBI:24579
IUPAC Name hex-1-ene
SMILES CCCCC=C

Specifications

Melting Point -140°C
Color Yellow
Boiling Point 64°C
Quantity 25 mL
UN Number 2370
Formula Weight 84.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexene
EINECSNumber (2)-0022&(2)-0031
RTECSNumber MP6670000
TSCA Yes