missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene 99.5+%, TCI America™

Supplier:  TCI America S03365ML

Catalog No. S03365ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Hexene [Standard Material for GC]
Colorless
C6H12
5 mL
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
CCCCC=C
84.162
CHEBI:24579
≥99.5% (GC)
592-41-6
63°C
MFCD00009505
2370
LIKMAJRDDDTEIG-UHFFFAOYSA-N
hex-1-ene
11597
84.16
Liquid

Chemical Identifiers

592-41-6
84.162
LIKMAJRDDDTEIG-UHFFFAOYSA-N
11597
hex-1-ene
C6H12
MFCD00009505
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
CHEBI:24579
CCCCC=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0022&(2)-0031

RTECSNumber : MP6670000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions