missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene, 99%, AcroSeal™

Catalog No. p-4229238
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120751000 100 mL AcroSeal™ Glass Bottle
AC120750010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120751000 Supplier Thermo Scientific Chemicals Supplier No. 120751000
Only null left

CAS: 592-41-6 | C6H12 | 84.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 592-41-6
Molecular Formula C6H12
Molecular Weight (g/mol) 84.15
MDL Number MFCD00009505
InChI Key LIKMAJRDDDTEIG-UHFFFAOYSA-N
Synonym 1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
PubChem CID 11597
ChEBI CHEBI:24579
IUPAC Name hex-1-ene
SMILES CCCCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -140.0°C
Color Colorless
Density 0.6780g/mL
Boiling Point 62.0°C to 65.0°C
Flash Point −26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3867 to 1.3887
Linear Formula CH3(CH2)3CH=CH2
Quantity 100 mL
Beilstein 01,215
Specific Gravity 0.678
Solubility Information Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether
Viscosity 0.34 mm2/s (40°C)
Formula Weight 84.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Hexene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Imm
EINECSNumber 209-753-1

RUO – Research Use Only