missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene, 98%

Catalog No. p-7024986
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB20271AE 100 mL
AAB20271AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB20271AE Supplier Thermo Scientific Chemicals Supplier No. B20271AE
Only null left

CAS: 592-41-6 | C6H12 | 84.162 g/mol

1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.

Solubility
Miscible with water, ethanol, ethyl ether, benzene and petroleum ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 592-41-6
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009505
InChI Key LIKMAJRDDDTEIG-UHFFFAOYSA-N
Synonym 1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
PubChem CID 11597
ChEBI CHEBI:24579
IUPAC Name hex-1-ene
SMILES CCCCC=C

Specifications

Melting Point -140°C
Density 0.673
Boiling Point 62°C to 64°C
Flash Point −25°C (−13°F)
Odor Mild
Refractive Index 1.388
Linear Formula CH3(CH2)3CH=CH2
Quantity 100 mL
UN Number UN2370
Beilstein 1209240
Solubility Information Miscible with water,ethanol,ethyl ether,benzene and petroleum ether.
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material 1-Hexene
Show More Show Less
Hazard Category H225-H304-H336
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-HEXENE
EINECSNumber 209-753-1
RTECSNumber MP6670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only