missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 1-Hexen-5-yne | |
| Yellow | |
| MFCD00060909 | |
| 3295 | |
| C=CCCC#C | |
| 80.13 | |
| 80.13 | |
| Liquid |
| 14548-31-3 | |
| C6H8 | |
| 5 g | |
| GHGNRQPVGKFJIR-UHFFFAOYSA-N | |
| hex-1-en-5-yne | |
| 518966 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 14548-31-3 | |
| 80.13 | |
| GHGNRQPVGKFJIR-UHFFFAOYSA-N | |
| hex-1-en-5-yne |
| C6H8 | |
| MFCD00060909 | |
| 518966 | |
| C=CCCC#C |
Safety and Handling
TSCA : No