missing translation for 'onlineSavingsMsg'
Learn More

1-Hexen-5-yne 97.0+%, TCI America™

Catalog No. H15411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15411G 1 g
H15415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15411G Supplier TCI America Supplier No. H15411G

Chemical Identifiers

CAS 14548-31-3
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00060909
InChI Key GHGNRQPVGKFJIR-UHFFFAOYSA-N
PubChem CID 518966
IUPAC Name hex-1-en-5-yne
SMILES C=CCCC#C

Specifications

Color Yellow
Quantity 1 g
UN Number 3295
Formula Weight 80.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexen-5-yne