missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexanol 98.0+%, TCI America™
$47.46 - $62.66
Chemical Identifiers
| CAS | 111-27-3 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.177 |
| MDL Number | MFCD00002982 |
| InChI Key | ZSIAUFGUXNUGDI-UHFFFAOYSA-N |
| Synonym | 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol |
| PubChem CID | 8103 |
| ChEBI | CHEBI:87393 |
| IUPAC Name | hexan-1-ol |
| SMILES | CCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H013025ML
|
TCI America
H013025ML |
25 mL |
Each for $47.46
|
|
|||||
|
H0130500ML
|
TCI America
H0130500ML |
500 mL |
Each for $62.66
|
|
|||||
Chemical Identifiers
| 111-27-3 | |
| 102.177 | |
| ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
| 8103 | |
| hexan-1-ol |
| C6H14O | |
| MFCD00002982 | |
| 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
| CHEBI:87393 | |
| CCCCCCO |
Specifications
| 111-27-3 | |
| Colorless | |
| C6H14O | |
| 25 mL | |
| 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
| CCCCCCO | |
| 102.177 | |
| CHEBI:87393 | |
| ≥98.0% (GC) | |
| 1-Hexanol |
| -45°C | |
| 157°C | |
| MFCD00002982 | |
| 2282 | |
| ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
| hexan-1-ol | |
| 8103 | |
| 102.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MQ4025000
TSCA : Yes