missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol 98.0+%, TCI America™

Catalog No. H0130500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0130500ML 500 mL
H013025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0130500ML Supplier TCI America Supplier No. H0130500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point -45°C
Color Colorless
Boiling Point 157°C
Quantity 500 mL
UN Number 2282
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexanol
EINECSNumber (2)-0217
RTECSNumber MQ4025000
TSCA Yes