missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A182320F

 View more versions of this product

Catalog No. AAA182320F



Description

Description

1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.

Solubility
Miscible with ethanol, acetone, chloroform, ether, benzene. Slightly miscible with carbon tetrachloride and water.

Notes
Incompatible with strong acids, acid chlorides, acid anhydrides and strong oxidizing agents.
Specifications

Specifications

1-Hexanol
-52°C
157°C
99%
1.418
MFCD00002982
UN2282
14,4697
Miscible with ethanol,acetone,chloroform,ether,benzene. Slightly miscible with carbon tetrachloride and water.
CCCCCCO
102.177
CHEBI:87393
99%
111-27-3
0.814
60°C (140°F)
C6H14O
CH3(CH2)5OH
2500 mL
969167
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
hexan-1-ol
8103
102.18

Chemical Identifiers

111-27-3
102.177
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
8103
hexan-1-ol
C6H14O
MFCD00002982
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
CHEBI:87393
CCCCCCO
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P362-P370+P378q-P374-P380-P501c

H226-H302+H312-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS

EINECSNumber : 203-852-3

RTECSNumber : MQ4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only