missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecyne, tech. 90%

Catalog No. AAA1136606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1136606 5 g
AAA1136614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1136606 Supplier Thermo Scientific Chemicals Supplier No. A1136606

CAS: 629-74-3 | C16H30 | 222.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-74-3
Molecular Formula C16H30
Molecular Weight (g/mol) 222.42
MDL Number MFCD00015084
InChI Key UCIDYSLOTJMRAM-UHFFFAOYSA-N
Synonym 1-hexadecyne, tetradecylacetylene, acmc-209nbc
PubChem CID 12396
IUPAC Name hexadec-1-yne
SMILES CCCCCCCCCCCCCCC#C

Specifications

Melting Point 13°C to 16°C
Density 0.79
Boiling Point 146°C to 148°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.442
Linear Formula CH3(CH2)13C≡CH
Quantity 5 g
Beilstein 1702804
Formula Weight 222.42
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Hexadecyne
Show More Show Less
EINECSNumber 211-106-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only