missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)pyridinium chloride monohydrate, 98%

Catalog No. aaa1349922
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAA1349922 100 g
AAA1349914 25 g
AAA1349936 500 g
3 options

Catalog No. AAA1349922

Supplier: Thermo Scientific Chemicals A1349922

Only null left

CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol

(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

Solubility
Soluble in water, alcohol, chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

Melting Point 83°C to 86°C
Quantity 100 g
UN Number UN2811
Beilstein 3578606
Merck Index 14,2032
Solubility Information Soluble in water, alcohol, chloroform
IUPAC Name 1-hexadecylpyridin-1-ium;chloride;hydrate
Formula Weight 358.02 (339.99 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1-Hexadecyl)pyridinium chloride monohydrate
Hazard Category H301-H315-H318-H330-H335
Hazard Statement GHS H Statement
H300-H330-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 204-593-9
RTECSNumber UU5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only