missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanesulfonyl chloride, 97+%

Catalog No. AAA1439609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1439609 10 g
AAA1439618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1439609 Supplier Thermo Scientific Chemicals Supplier No. A1439609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38775-38-1 | C16H33ClO2S | 324.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38775-38-1
Molecular Formula C16H33ClO2S
Molecular Weight (g/mol) 324.95
MDL Number MFCD00007459
InChI Key YOSVFFVBSPQTTP-UHFFFAOYSA-N
Synonym 1-hexadecanesulfonyl chloride, hexadecylsulfonyl chloride, 1-hexadecanesulfonylchloride, hexadecane-1-sulphonyl chloride, acmc-20aouo, hexadecanesulfonylchloride, hexadecanesulfonyl chloride, 1-hexadecylsulfonyl chloride, cetyl sulphonic acid chloride, 1-hexadecanesulphonyl chloride
PubChem CID 96923
IUPAC Name hexadecane-1-sulfonyl chloride
SMILES CCCCCCCCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 56°C to 58°C
Quantity 10 g
UN Number UN3261
Beilstein 1789245
Sensitivity Moisture sensitive
Formula Weight 324.96
Percent Purity 98%
Chemical Name or Material 1-Hexadecanesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 254-123-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only