missing translation for 'onlineSavingsMsg'
Learn More

1-Hepten-4-ol, 97+%

Catalog No. p-7025215
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2059103 1 g
AAB2059106 5 g
AAB2059114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2059103 Supplier Thermo Scientific Chemicals Supplier No. B2059103
Only null left

CAS: 3521-91-3 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3521-91-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00021933
InChI Key AQTUHJABKZECGA-UHFFFAOYNA-N
Synonym 1-hepten-4-ol, acmc-20aouq
PubChem CID 19040
IUPAC Name hept-1-en-4-ol
SMILES CCCC(O)CC=C

Specifications

Refractive Index 1.435
Quantity 1 g
UN Number UN1987
Formula Weight 114.18
Percent Purity 98%
Chemical Name or Material 1-Hepten-4-ol
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 222-535-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only