missing translation for 'onlineSavingsMsg'
Learn More

1-Hepten-3-ol, 98%

Catalog No. AAA1465109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1465109 10 g
AAA1465118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1465109 Supplier Thermo Scientific Chemicals Supplier No. A1465109

CAS: 4938-52-7 | C7H14O | 114.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4938-52-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00021940
InChI Key PZKFYTOLVRCMOA-UHFFFAOYSA-N
Synonym 1-hepten-3-ol, heptene-1-ol-3, butyl vinyl carbinol, acmc-1aoyf, 1-hepten-3-ol, analytical standard, 1-hepten-3-ol, fg, 1-hepten-3-ol, purum gc
PubChem CID 21057
IUPAC Name hept-1-en-3-ol
SMILES CCCCC(C=C)O

Specifications

Density 0.836
Boiling Point 154°C to 156°C
Flash Point 54°C (129°F)
Refractive Index 1.433
Quantity 10 g
UN Number UN1987
Beilstein 1720340
Formula Weight 114.18
Percent Purity 98%
Chemical Name or Material 1-Hepten-3-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 225-579-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only