missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, 99%

Catalog No. AAA12793AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12793AE 100 mL
AAA12793AP 500 mL
AAA127930F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12793AE Supplier Thermo Scientific Chemicals Supplier No. A12793AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-70-6 | C7H16O | 116.20 g/mol

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Melting Point -34°C
Density 0.82
Boiling Point 175°C to 176°C
Flash Point 73°C (163°F)
Odor Characteristic
Assay Percent Range 99%
Refractive Index 1.424
Linear Formula CH3(CH2)6OH
Quantity 100 mL
Beilstein 1731686
Merck Index 14,4661
Solubility Information Soluble in water 1.7g/L at 25°C.
Formula Weight 116.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Heptanol
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H311-H227-H302-H332-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 203-897-9
RTECSNumber MK0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only