missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, 99%

CAS: 111-70-6 | C7H16O | 116.20 g/mol

Supplier:  Thermo Scientific Chemicals A127930F

Quantity 2500 mL

 View more versions of this product

Catalog No. AAA127930F


Only null left

Description

Description

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
Specifications

Chemical Identifiers

111-70-6
116.20
BBMCTIGTTCKYKF-UHFFFAOYSA-N
8129
CCCCCCCO
C7H16O
MFCD00002986
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
heptan-1-ol

Specifications

1-Heptanol
-34°C
175°C to 176°C
Characteristic
C7H16O
CH3(CH2)6OH
MFCD00002986
14,4661
Soluble in water 1.7g/L at 25°C.
CCCCCCCO
116.20
116.2
Liquid
111-70-6
0.82
73°C (163°F)
99%
1.424
2500 mL
1731686
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
BBMCTIGTTCKYKF-UHFFFAOYSA-N
heptan-1-ol
8129
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H227-H302-H332-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P264b-P280-P305+P351+P338-P370+P378q-P501c

H227-H319

EINECSNumber : 203-897-9

RTECSNumber : MK0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions