missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanethiol 98.0+%, TCI America™

Catalog No. H002925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H002925G 25 g
H002910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H002925G Supplier TCI America Supplier No. H002925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1639-09-4
Molecular Formula C7H16S
Molecular Weight (g/mol) 132.265
MDL Number MFCD00004911
InChI Key VPIAKHNXCOTPAY-UHFFFAOYSA-N
Synonym 1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
PubChem CID 15422
IUPAC Name heptane-1-thiol
SMILES CCCCCCCS

Specifications

Melting Point -43°C
Color Colorless
Boiling Point 177°C
Quantity 25 g
UN Number 3336
Formula Weight 132.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Heptanethiol
EINECSNumber (2)-0464
RTECSNumber MJ1400000
TSCA Yes